To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:2" Cardboard Freezer Boxes
- Cat. no.:102093-720
- Supplier No.:61615-65
Specifications
About this item
For use with stainless steel freezer racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes.
- Box footprint is 133×133 mm
Select a box and a partition that ft the size and number of vials for your needs. Cardboard boxes can be labelled with Cryo-Tags for easy identification.