Spécifications
- Description:Spinbar™ Restrainer
- Cat. no.:102091-104
- Supplier No.:64526
Spécifications
A propos de cet article
The Spinbar Restrainer fits in one hand and when placed alongside the outer wall of a mixing vessel, it readily attracts and holds stirring bar in place, without contaminating solution being stirred.
As vessel is drained, the stirring bar is securely retrieved from vacant vessel. The Spinbar Restrainer id molded of easy to clean high impact polystyrene. It contains a powerful magnet that attracts and holds magnetic stirring bars up to 80 mm (3") long.