ContactUsLinkComponent
Restrainer Spinbar
SpinBar® Restrainer, Electron Microscopy Sciences
# de catalogue 102091-104
Restrainer Spinbar
SpinBar® Restrainer, Electron Microscopy Sciences
# de catalogue 102091-104
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Spinbar™ Restrainer
  • Cat. no.:
    102091-104
  • Supplier No.:
    64526
Spécifications
A propos de cet article
The Spinbar Restrainer fits in one hand and when placed alongside the outer wall of a mixing vessel, it readily attracts and holds stirring bar in place, without contaminating solution being stirred.
As vessel is drained, the stirring bar is securely retrieved from vacant vessel. The Spinbar Restrainer id molded of easy to clean high impact polystyrene. It contains a powerful magnet that attracts and holds magnetic stirring bars up to 80 mm (3") long.
Customers Who Bought This Also Bought