ContactUsLinkComponent
Staining Kit Live/Dead Cells
Cell Viability and Proliferation Assays, Biotium
# de catalogue 89260-208
Fournisseur:  Biotium
Numéro CAS:  
Cell Viability and Proliferation Assays, Biotium
# de catalogue 89260-208
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Viability/Cytotoxicity Assay Kit for Animal Live and Dead Cells
  • Cat. no.:
    89260-208
  • Kit Size:
    150 assays
  • Features:
    Green/red fluorescent staining of live and dead cells
  • Includes:
    1
  • Supplier No.:
    30002-T
Spécifications
A propos de cet article
Biotium offers cell proliferation assays to quantitate live or dead cells by absorbance, fluorescence, or luminescence.
  • Aassay employs two probes that detect intracellular esterase activity in live cells and compromised plasma membrane integrity in dead cells
Calcein-AM and ViaFluor™ CFSE are initially colorless compounds that are hydrolyzed by cellular esterases to label the cytoplasm with green fluorescence. Calcein-AM and ViaFluor™ CFSE can be used to quantitate viable cells by fluorescence microplate reader. ViaFluor™ CFSE stably labels cells and also can be used to track cell divisions over multiple generations by flow cytometry analysis in the FITC channel. The Cytotoxicity and Viability Assay pairs calcein-AM with the red fluorescent vital dye ethidium homodimer III for quantitation of live and dead cells.
The ATP-Glo cell viability assay uses Firefly luciferase lumenescence to measure cellular ATP levels. ATP-Glo is highly sensitive, and can detect as little as one cell or 0.01 picomole of ATP, with signal linearity for ATP detection within 6 orders of magnitude. ATP-Glo is a flash-type luminescence assay (signal is stable for ~ 1 minute), for use with a single read luminometer or luminescence plate reader with sample injector.
MTT and XTT are reduced by viable cells to form colored formazin salts, which can be measured by absorbance. MTT requires cell lysis prior to measurement, while XTT does not. Resazurin is reduced by viable cells from a dark blue colored compound to a red fluorescent compound, which can be measured by fluroescence or absorbance.
Ordering information: Kit size based on 96-well assay. CFDA-SE assay number varies by application.
Customers Who Bought This Also Bought