VWR® Precision Tweezers
# de catalogue 89259-984
Fournisseur: VWR International
Spécifications
- Length:120 mm
- Description:Precision Tweezers, Style No. 3
- Material:Stainless steel
- Cat. no.:89259-984
Spécifications
A propos de cet article
VWR® Precision Tweezers are stainless steel with very sharp, fine points.
- Anti-magnetic properties
- Resistant to acids
- Tweezers feature fine points
Precision tweezers are available in a range of lengths and order sizes, from individual to bulk.
Customers Who Bought This Also Bought