ContactUsLinkComponent
Operating Scissors
VWR® Operating Scissors
# de catalogue 89259-980
Fournisseur:  VWR International
Operating Scissors
VWR® Operating Scissors
# de catalogue 89259-980
Fournisseur:  VWR International
Spécifications
  • Material:
    Stainless steel
  • Cat. no.:
    89259-980
  • Length:
    114 mm (41/2")
Spécifications
A propos de cet article
VWR® Operating Scissors are made of stainless steel with straight, blunt tips.
  • Stainless steel construction
  • Straight, blunt tips
Operating scissors are available in a range of lengths and order sizes, from individual to bulk.
Customers Who Bought This Also Bought