VWR® Laboratory Spoon
# de catalogue 89259-966
Fournisseur: VWR International
Spécifications
- Length:229 mm (9")
- Material:Stainless steel
- Cat. no.:89259-966
Spécifications
A propos de cet article
VWR® Lab Spoons are stainless steel and useful in various applications.
- Stainless steel construction
Lab spoons are available in different lengths and in individual or bulk packages.
Customers Who Bought This Also Bought