VWR® Filter Forceps
# de catalogue 89259-954
Fournisseur: VWR International
Spécifications
- Length:114 mm (41/2")
- Material:Stainless steel
- Cat. no.:89259-954
Spécifications
A propos de cet article
VWR® Filter Forceps are stainless steel with black polypropylene non-slip finger grips.
- Black polypropylene fabrication
- Non-slip finger grips
The tips are flat for handling membranes.
Customers Who Bought This Also Bought