VWR® Cushing Forceps
# de catalogue 89259-952
Fournisseur: VWR International
Spécifications
- Length:178 mm (7")
- Material:Stainless steel
- Cat. no.:89259-952
Spécifications
A propos de cet article
VWR® Cushing Forceps are stainless steel with straight, serrated tips.
- Stainless steel construction
- Straight, serrated tips
They feature a corrugated handle and a guide pin.
Customers Who Bought This Also Bought