VWR® Dissecting Forceps with Guide Pin
Catalog # 89259-946
Supplier: VWR International
Specifications
- Description:Dissecting Forceps with Curved Tips
- Length:114 mm (4½")
- Material:Stainless steel
Specifications
About this item
VWR® Dissecting Forceps are stainless steel with fine-point serrated tips and a guide pin.
- Stainless steel construction
- Fine-pointed serrated tips
Dissecting forceps are used for a variety of applications and are available in different lengths.
Frequently Bought Together