VWR® Dissecting Forceps with Guide Pin
# de catalogue 89259-944
Fournisseur: VWR International
Spécifications
- Length:114 mm (41/2")
- Description:Dissecting Forceps with Straight Tips
- Material:Stainless steel
- Cat. no.:89259-944
Spécifications
A propos de cet article
VWR® Dissecting Forceps are stainless steel with fine-point serrated tips and a guide pin.
- Stainless steel construction
- Fine-pointed serrated tips
Dissecting forceps are used for a variety of applications and are available in different lengths.
Customers Who Bought This Also Bought