VWR® Reverse Action Forceps
# de catalogue 89259-942
Fournisseur: VWR International
Spécifications
- Length:127 mm (5")
- Material:Stainless steel
- Cat. no.:89259-942
Spécifications
A propos de cet article
VWR® Reverse Action Forceps open with finger pressure.
- Stainless steel construction
- Finger-pressure open action
- Sharp, smooth points
They are stainless steel with sharp, smooth points.
Customers Who Bought This Also Bought