VWR® Micro Spatulas, Double-Ended
# de catalogue 89259-890
Fournisseur: VWR International
Spécifications
- Description:Micro spatula with bent, sharpened tip and V-shaped sharpened spoon at other end
- Cat. no.:89259-890
- Blade Length:170 mm
- Blade Width:2 mm
Spécifications
A propos de cet article
These VWR® Micro spatulas are double-ended and stainless steel.
- Stainless steel construction
- Double-ended design
Spatula has a curved spatula. Spatula is sharpened at the tip for cutting and separating and has a small knob at other end for crushing crystals.
Customers Who Bought This Also Bought