VWR® Neoprene Crucible Holders
# de catalogue 89259-858
Fournisseur: VWR International
Spécifications
- Description:Neoprene Crucible Holder, Size 1, 20 mm height, 10 mm bottom O.D., 22 mm top O.D.
- Cat. no.:89259-858
Spécifications
A propos de cet article
These neoprene crucible holders are tapered, collar-type holders.
- Used alone or with adjacent sizes
They support funnels, crucibles, and other equipment in vacuum-type filtration assemblies. They feature a positive, but easy-to-release gas-tight seal.
Customers Who Bought This Also Bought