ContactUsLinkComponent
AccuStart II GelTrack PCR SuperMix, 500R
AccuStart™ II GelTrack PCR SuperMix, QuantaBio
Catalog # CA89235-014
Supplier:  Quantabio
CAS Number:  
AccuStart™ II GelTrack PCR SuperMix, QuantaBio
Catalog # CA89235-014
Supplier:  Quantabio
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    AccuStart™ II GelTrack PCR SuperMix
  • Size:
    500 rxn (of 25 µl rxn)
  • Cat. no.:
    CA89235-014
  • Supplier No.:
    95136-500
Specifications
About this item
AccuStart II GelTrack PCR SuperMix is a 2X concentrated, ready-to-use reaction cocktail for routine PCR amplification of DNA fragments up to 4 kb. It contains premixed loading dye to streamline gel electrophoresis and all components, except primers and template.
  • High yield; high sensitivity
  • Precise amplification
  • Superior reproducibility and precision
  • Convenient reaction assembly at room temperature
AccuStart II PCR SuperMix simplifies reaction assembly, improves assay reproducibility, and reduces the risk of contamination. A key component is AccuStart™ II Taq DNA polymerase which contains monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation (1 minute at 94 °C), the antibodies denature irreversibly, releasing fully active, unmodified Taq DNA polymerase. This enables specific and efficient primer extension with the convenience of room temperature reaction assembly.
Store components in a constant temperature freezer at −25 to −15 °C upon receipt. Repeated freezing and thawing does not impair product performance. For lot specific expiry date, refer to package label, Certificate of Analysis or Product Specification Form.
AccuStart II GelTrack PCR SuperMix is a 2X concentrated, ready-to-use reaction cocktail for routine PCR amplification of DNA fragments up to 4 kb. It contains premixed loading dye to streamline gel electrophoresis and all components, except primers and template.
Ordering information: AccuStart™ II GelTrack PCR Supermix contains 2X reaction buffer containing 3 mM MgCl₂, 0.4 mM each dNTP (dATP, dCTP, dGTP, dTTP), Accustart II Taq DNA Polymerase and GelTrack loading dye and stabilizers. For research only. Not intended for any animal or human therapeutic or diagnostic use. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Customers Who Bought This Also Bought