ContactUsLinkComponent
SNAP CAP, PTFE/SIL/PTFE, BLU
WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
# de catalogue 89235-568
Fournisseur:  DWK Life Sciences
Small Business Enterprise
SNAP CAP, PTFE/SIL/PTFE, BLU
WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
# de catalogue 89235-568
Fournisseur:  DWK Life Sciences
Small Business Enterprise
Spécifications
  • Septum:
    PTFE/silicone/PTFE
  • Description:
    Snap cap for E-Z Vial® with snap ring top
  • Cat. no.:
    89235-568
  • Cap Color:
    Light blue
  • Supplier No.:
    242772-01
Spécifications
A propos de cet article
These snap-top caps eliminate the need for crimping and decapping tools.
  • Manufactured from PE
  • Fits the WHEATON E-Z Vials® with snap ring top
Available in multiple colors and septa types.
Customers Who Bought This Also Bought