WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
# de catalogue 89235-568
Fournisseur: DWK Life Sciences
Small Business Enterprise
Spécifications
- Septum:PTFE/silicone/PTFE
- Description:Snap cap for E-Z Vial® with snap ring top
- Cat. no.:89235-568
- Cap Color:Light blue
- Supplier No.:242772-01
Spécifications
A propos de cet article
These snap-top caps eliminate the need for crimping and decapping tools.
- Manufactured from PE
- Fits the WHEATON E-Z Vials® with snap ring top
Available in multiple colors and septa types.
Customers Who Bought This Also Bought