VWR® Basic Apron, Yellow
# de catalogue 89232-928
Fournisseur: VWR International
Spécifications
- Length:145 cm (57")
- Color:Yellow
- Description:Basic Apron
- Cat. no.:89232-928
Spécifications
A propos de cet article
VWR® basic aprons are designed to provide protection against light splash and provide expanded coverage versus traditional aprons.
- Extra long (57")
- Elastic wrists, open back, bound seams
Aprons are extra long (57") to provide additional coverage below the knee. They feature elastic wrists, open back, and bound seams for maximum protection.
Customers Who Bought This Also Bought