L(+)-Ascorbic acid 99.0-100.5% USP, Macron Fine Chemicals™
# de catalogue CAMK185210
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 50-81-7
MOC : RéglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber, wide mouth
- Size:500 g
- Cat. no.:CAMK185210
- Supplier No.:1852-10
Spécifications
Résultats des tests de spécification
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Appearance (fine, white to slightly yellow crystals):Passes Test
- USP - Assay (C₆H₈O₆):99.0 - 100.5 %
- USP - Specific Rotation [Å]²⁵^D (+):20.5 - 21.5 Degree
- USP - Residue on Ignition:≤0.1 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- Residual Solvents - Only Class 2 Solvents (Methanol) and Class 3:
- Solvents are likely to be present. Residual Class 2 Solvents are below:
- Option 1 and the residual concentration of all Class 3 Solvents is:
- <0.5%.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Résultats des tests de spécification
A propos de cet article
Synonymes: Ascorbic Acid,(R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one
Point d'ébullition:553 °C (1013 hPa)
Point de Fusion:190…192 °C
Densité:1.694 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 553 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 190…192 °C
:Flash Pt:
:Flash Pt:
:M.W. 176.13 g/mol
Index Merck:13,00837
:L(+)-Ascorbic acid
Température d'entreposage:Ambient
EINECS:200-066-2
:exempt
:M.O.C. Category R
Customers Who Bought This Also Bought