ContactUsLinkComponent
GRAM STAIN CONTROL SLIDES PK50
Parasitology Control Slides, Azer Scientific
# de catalogue 89233-248
Fournisseur:  Azer Scientific Inc.
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
GRAM STAIN CONTROL SLIDES PK50
Parasitology Control Slides, Azer Scientific
# de catalogue 89233-248
Fournisseur:  Azer Scientific Inc.
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Gram stain control slides
  • Supplier No.:
    ESCS-141
Spécifications
A propos de cet article
These parasitology control slides are broken into types.
  • Space on each side for writing
  • Control organisms: Staph. aureus and E. coli
  • For placement of clinical smears on the same slide with control organisms
Gram-stain control slides are specially designed so clinical smears can be placed on the same slide with the control organism.
Space is provided on each slide for writing.
Packaging: 50 per box
Customers Who Bought This Also Bought