Spécifications
- Description:Gram stain control slides
- Supplier No.:ESCS-141
Spécifications
A propos de cet article
These parasitology control slides are broken into types.
- Space on each side for writing
- Control organisms: Staph. aureus and E. coli
- For placement of clinical smears on the same slide with control organisms
Gram-stain control slides are specially designed so clinical smears can be placed on the same slide with the control organism.
Space is provided on each slide for writing.