Acetone ≥99.5% ACS
# de catalogue CA71006-476
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 67-64-1
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Metal can
- Size:20 L
- Cat. no.:CA71006-476
- Supplier No.:00884-700
Spécifications
Résultats des tests de spécification
- Assay (CH3)2CO:99.5% min.
- Color (APHA):10 max.
- Residue after evaporation:0.001% max.
- Solubility in water:Passes Test
- Titrable acid:0.0003 meq/gmax.
- Titrable base:0.0006 meq/g max.
- Aldehyde (as HCHO):0.002% max.
- Isopropyl Alcohol:0.05% max.
- Methanol:0.05% max.
- Substances reducing KMnO4:Passes Test
- Water (H2O):0.5% max.
- Acetone ≥99.5% ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: 2-Propanone,DMK,Propanone
Point d'ébullition:56.2 °C (1013 hPa)
Point de Fusion:–95.4 °C
Densité:0.792 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
Point d'éclair:<–20 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 56.2 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –95.4 °C
:Flash Pt:
:Flash Pt: Max. -20 °C (closed cup)
:M.W. 58.08 g/mol
Index Merck:13,00067
EINECS:200-662-2
:-95.4
Température d'entreposage:Ambient
ONU:1090
:01-2119471330-49
:3
:II
:<–20 °C
:GHS02
:GHS07
Customers Who Bought This Also Bought