Order Entry
ContactUsLinkComponent
Vwr Bottle Wm Hdpe 30Ml - Bulk
VWR® Laboratory Bottles, HDPE
Catalog # 89221-658
Supplier:  VWR International
Vwr Bottle Wm Hdpe 30Ml - Bulk
VWR® Laboratory Bottles, HDPE
Catalog # 89221-658
Supplier:  VWR International

Specifications

  • Cap size:
    28-415
  • Capacity:
    30 ml (1 oz.)
  • Description:
    Wide mouth bottles
  • Cat. no.:
    89221-658

Specifications

About this item

Translucent, rigid bottles are for general laboratory use and for storing, shipping, and packing liquids.

  • Durable for long-term use
  • High-density polyethylene construction
  • Translucent siding

Can be stored in freezer to −100 °C (−148 °F). Excellent chemical resistance to most acids, bases, and alcohols. Can be used with most corrosives.

Delivery: Polypropylene screw closure included.
Customers Who Bought This Also Bought