Spécifications
- Description:Hamilton Model 1702 Replacement Plunger
- Cat. no.:89221-376
- Supplier No.:13269
- Termination:TLLX, DX, C
Spécifications
A propos de cet article
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
These replacement plungers are for Hamilton 1700 series Gastight® syringes ranging in volume from 10 to 500 µl.
Each is manufactured to achieve the highest level of accuracy and precision possible.