ContactUsLinkComponent
Caps, AQR, autosampler
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
# de catalogue 89220-006
Caps, AQR, autosampler
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
# de catalogue 89220-006
Spécifications
  • Cap Size:
    11 mm
  • Cap Type:
    Snap cap
  • Septum:
    Fitted Ultra Pure Clear Silicone/PTFE
  • Material:
    PE
  • Cat. no.:
    89220-006
  • Cap Color:
    Light Blue
  • Supplier No.:
    9509C-10CB