AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
# de catalogue 89220-006
Fournisseur: MicroSolv Technology Corp
Spécifications
- Cap Size:11 mm
- Cap Type:Snap cap
- Septum:Fitted Ultra Pure Clear Silicone/PTFE
- Material:PE
- Cat. no.:89220-006
- Cap Color:Light Blue
- Supplier No.:9509C-10CB