Specifications
- Type:Sterile
- Description:Sterile Syringe Caps
- Cat. no.:89219-284
- Packaging:100/box
- For Use With:Laboratory and Veterinary Use Only
- Supplier No.:STC
Specifications
About this item
Cap off any of your Luer slip or Luer lock syringes with these cost-efficient, sterile caps.
- Sterile
- Fits all Luer lock and Luer slip syringes
- Polypropylene
For capping off and sealing syringes, these sterile, polypropylene syringe caps fit all Luer slip and Luer lock syringes.