Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
# de catalogue JT9264-3
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 75-09-2
MOC : With Information"Certificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 L
- Supplier No.:9264-03
Spécifications
Résultats des tests de spécification
- For Laboratory,Research,or Manufacturing Use:
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
- Assay (CH₂Cl₂) (by GC, exclusive of preservative, corrected for water):≥99.8 %
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Titrable Acid (µeq/g):≤0.3
- Chloride (Cl):≤10 ppm
- Water (by KF, coulometric):≤0.02 %
- Contains a methanol-alkene preservative system.:
Résultats des tests de spécification
A propos de cet article
Synonymes: Methylene chloride
Point d'ébullition:39.8 °C (1013 hPa)
Point de Fusion:–95 °C
Densité:1.322 g/cm³ (20 °C)
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Packaged under inert gas.
:Stabilised with a methanol-alkene preservative system
:Boiling Pt:
:Boiling Pt: 39.8 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –95 °C
:Flash Pt:
:Flash Pt:
:M.W. 84.93 g/mol
Index Merck:12,06140
:Dichloromethane
Température d'entreposage:Ambient
EINECS:200-838-9
:-95
ONU:1593
:6.1
:III
:GHS08
:GHS07
:Methanol-alkene
:M.O.C. Category NI
Customers Who Bought This Also Bought