Methanol ≥99.9%, Semi Grade for the electronics industry, VWR Chemicals BDH®
# de catalogue BDH2029-1GLP
Fournisseur: BDH
Numéro CAS: 67-56-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:HDPE bottle
- Size:1 gal.
- Supplier No.:10705S1G41BD
Spécifications
Résultats des tests de spécification
- Assay (CH3OH):99.9% Min.
- Color (APHA):10 APHA Max.
- Acidity:0.3 µeq/g Max.
- Alkalinity:0.1 µeq/g Max.
- Residue after Evaporation:5 ppm Max.
- Water (H2O):0.05% Max.
- Solubility in Water:To Pass Test
- Chloride (Cl):200 ppb Max.
- Phosphate (PO4):500 ppb Max.
- Arsenic and Antimony:10 ppb Max.
- Al Impurities:100 ppb Max.
- B Impurities:100 ppb Max.
- Ca Impurities:100 ppb Max.
- Cr Impurities:100 ppb Max.
- Cu Impurities:100 ppb Max.
- Au Impurities:100 ppb Max.
- Fe Impurities:100 ppb Max.
- Mg Impurities:100 ppb Max.
- Mn Impurities:100 ppb Max.
- Ni Impurities:100 ppb Max.
- K Impurities:100 ppb Max.
- Na Impurities:100 ppb Max.
- Pb Impurities:100 ppb Max.
- Sn Impurities:100 ppb Max.
- Ti Impurities:100 ppb Max.
- Zn Impurities:100 ppb Max.
- Particles ≥1.0 µm:10/mL Max.
- VWR Chemicals BDH®, Methanol ≥99.9%, Semi Grade for the electronics industry:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydroxy methane,Carbinol,Wood alcohol
Point d'ébullition:64.6 °C (1013 hPa)
Point de Fusion:–98 °C
Densité:0.7918 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
Point d'éclair:11 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 64.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 11 °C (closed cup)
:M.W. 32.04 g/mol
Index Merck:13,05984
EINECS:200-659-6
:-98
Température d'entreposage:Ambient
ONU:1230
:01-2119433307-44
:3
:II
:11
:GHS02
:GHS06
:GHS08
Customers Who Bought This Also Bought