Spécifications
- Color:Clear
- Description:Frozen Section Compound
- Cat. no.:95057-838
Spécifications
A propos de cet article
Frozen section compound is a fast-acting, water-soluble embedding compound designed for use in frozen sectioning.
Frozen section compound is a fast-acting, water-soluble embedding compound designed for use in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. At a working temperature of –20°C (–4°F), it provides optimal sectioning consistency with minimal section curling. Each bottle features an extended length tip for increased accuracy in dispensing. Highly recommended for surgical pathology laboratories. Available clear or in a light blue format for better visualization of small specimens. Size: 118mL (4oz.).
- Water-soluble embedding compound
- Clear siding for quick inspection
- Minimal section curling
The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. At a working temperature of -20°C (-4°F), it provides optimal sectioning consistency with minimal section curling. Each bottle features an extended length tip for increased accuracy in dispensing. Highly recommended for surgical pathology laboratories. Available in a clear format for better visualization of small specimens. Size: 118 mL (4 oz.).