ContactUsLinkComponent
Vwr Clear Frozen Section Compound
VWR® Premium Frozen Section Compound
# de catalogue 95057-838
Fournisseur:  VWR International
Product available on GSA Advantage®
Numéro CAS:  
Vwr Clear Frozen Section Compound
VWR® Premium Frozen Section Compound
# de catalogue 95057-838
Fournisseur:  VWR International
Product available on GSA Advantage®
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Clear
  • Description:
    Frozen Section Compound
  • Cat. no.:
    95057-838
Spécifications
A propos de cet article
Frozen section compound is a fast-acting, water-soluble embedding compound designed for use in frozen sectioning.
Frozen section compound is a fast-acting, water-soluble embedding compound designed for use in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. At a working temperature of –20°C (–4°F), it provides optimal sectioning consistency with minimal section curling. Each bottle features an extended length tip for increased accuracy in dispensing. Highly recommended for surgical pathology laboratories. Available clear or in a light blue format for better visualization of small specimens. Size: 118mL (4oz.).
  • Water-soluble embedding compound
  • Clear siding for quick inspection
  • Minimal section curling
The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. At a working temperature of -20°C (-4°F), it provides optimal sectioning consistency with minimal section curling. Each bottle features an extended length tip for increased accuracy in dispensing. Highly recommended for surgical pathology laboratories. Available in a clear format for better visualization of small specimens. Size: 118 mL (4 oz.).
Customers Who Bought This Also Bought