Order Entry
ContactUsLinkComponent
 
VACUETTE® Serum Clot Activator Tubes, Greiner Bio-One
  95057-389
Product available on GSA Advantage®
Blood Collection Tube 5 ml Serum with gel separator pk50
VACUETTE® Serum Clot Activator Tubes, Greiner Bio-One
  95057-389
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

 

  • Ø:
    13 mm
  • Volume:
    5 ml
  • Description:
    Z Serum Clot Activator Tube with Gel Separator
  • Length:
    100 mm
  • Cat. no.:
    95057-389
  • Tube Dimensions:
    13 x 100 mm
  • Ring Color:
    Yellow
  • Cap Color:
    Red
  • Supplier No.:
    456073

 

 

Tubes are designed for routine use in immunohematology testing and viral marker testing in screening and clinical laboratories.

  • Gel forms stable barrier between serum and blood cells
  • Tubes are effective in therapeutic drug monitoring testing

The inner wall of each serum tube is coated with microscopic silica particles which activate the coagulation process when tubes are inverted

VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with non-ridged pull caps that feature an exterior color-coded ring to indicate either the additive used or draw volume. A black ring indicates standard tubes, a yellow/gold ring indicates the inclusion of a separation gel, and a white ring indicates a tube designed to accommodate pediatric draw.

Tubes equipped with a gel separator contain a barrier gel in the base of the tube. During centrifugation, this gel forms a stable barrier between the serum and the blood cells, separating the serum from fibrin and cells. These tubes are particularly effective in therapeutic drug monitoring testing. Standard tubes are also available with no additive.

Customers Who Bought This Also Bought