ContactUsLinkComponent
VWR COVERALL TYVEK BOOT XL CS25
VWR® Signature™ Coveralls made with DuPont™ Tyvek® IsoClean® Material
# de catalogue 80200-624
Fournisseur:  VWR International
VWR COVERALL TYVEK BOOT XL CS25
VWR® Signature™ Coveralls made with DuPont™ Tyvek® IsoClean® Material
# de catalogue 80200-624
Fournisseur:  VWR International
Spécifications
  • Clothing Size:
    XL
  • Garment application:
    Laboratory
  • Color:
    White
  • Description:
    Coveralls with attached boots
  • Cat. no.:
    80200-624
Spécifications
A propos de cet article
Protective, durable, and comfortable coveralls provide an excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
All coveralls feature serged seams, laydown collar, front zipper closure, and elastic wrists. Available with elastic ankles, with attached Gripper™ boots, or with attached boots and hood. Boot attachments combine an embossed PVC exterior sole with a coarsely textured interior to ensure a secure, nonslip fit and feel.
  • Protective, durable, and comfortable
  • Excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
  • Chemically and biologically inert, low linting, and static free until laundered
  • Serged seams, laydown collar, front zipper closure, and elastic wrists
  • Available with elastic ankles, with attached Gripper™ boots, or with attached boots and hood
  • Boot attachments combine embossed PVC exterior sole with textured interior to ensure secure, nonslip fit and feel
Protective, durable, and comfortable coveralls provide an excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
All coveralls feature serged seams, laydown collar, front zipper closure, and elastic wrists. Available with elastic ankles, with attached Gripper™ boots, or with attached boots and hood. Boot attachments combine an embossed PVC exterior sole with a coarsely textured interior to ensure a secure, nonslip fit and feel.
Garments are chemically and biologically inert, low-linting, and static-free until laundered.
Customers Who Bought This Also Bought