Lens Cleaning Towelettes, C-Clear
# de catalogue 11200-007
Fournisseur: C-Clear
Spécifications
- Description:Packed Towelettes
- Cat. no.:11200-007
- Packaging:100/Box
- Supplier No.:70
Spécifications
A propos de cet article
Cardboard dispenser holds 100 individually wrapped 12.7×20.3 cm (5×8") towelettes that are treated with anti-fog/anti-static formula.
- Sturdy corrugated cardboard dispenser
- Pull-out bottom for easy dispensing
- Built-in tab for hanging
- Maintain eyewear properly with these premoistened towelettes
- Ideal for cleaning glass, plastic, and polycarbonate lenses
May also be used on side shields, face shields, goggles, respirators, CRT screens, and microscopes.
Ordering information: Bulk packed towelettes are supplied without cardboard dispensers.
Customers Who Bought This Also Bought