ContactUsLinkComponent
Blood Tube Holder Quickshield pk50
VACUETTE® Specimen Collection System, Greiner Bio-One
# de catalogue 95057-343
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
Blood Tube Holder Quickshield pk50
VACUETTE® Specimen Collection System, Greiner Bio-One
# de catalogue 95057-343
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Safety Holder
  • Cat. no.:
    95057-343
  • Supplier No.:
    450230
Spécifications
A propos de cet article
The completely assembled VACUETTE® Specimen Collection System is equipped with a rotational protective shield that can be activated with the aid of a solid surface.
  • Rotational protective shield prevents needlestick injuries
  • Full coverage through rotation provides extra user comfort
  • Designed for use with VACUETTE® multiple-sample drawing needles
The protective shield is attached to the holder and, once activated, keeps the needle contained to help prevent needlestick injuries. The rotational shield can rotate 360° to provide user comfort. Holders are designed exclusively for use with VACUETTE® multiple-sample drawing needles or VACUETTE® VISIO PLUS needles. The Specimen Collection System combines a safety tube holder and a VACUETTE® VISIO PLUS needle.
Customers Who Bought This Also Bought