Order Entry
Canada
ContactUsLinkComponent
Fendall® Saline Eyewash Bottles, Honeywell Safety
Catalogue # 56611-118
undefined
Fendall® Saline Eyewash Bottles, Honeywell Safety
Catalogue # 56611-118
Supplier Number:  32-000462-0000-H5

Spécifications

  • Description:
    Personal eyewash solution bottle
  • Size:
    118 ml (3.9 oz.)
  • Cat. no.:
    56611-118
  • Supplier No.:
    32-000452-0000

Spécifications

À propos de cet article

This bottle contains a sterile, buffered saline solution for emergency eye care.

  • Expiry dates and lot numbers for end-user protection
  • User-controlled volume delivery: one, four, and eight-ounce products

It delivers drops or an irrigating stream of solution, depending on the squeeze pressure applied to the bottle.

Each bottle features extended flow openings and twist-off tops for easy access in an emergency.

Frequently Bought Together