Hydrogen peroxide solution 30%, AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
# de catalogue MKV34004
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 7722-84-1
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:PE bottle
- Size:500 ml
- Cat. no.:MKV34004
- Supplier No.:V340-04
Spécifications
Résultats des tests de spécification
- ACS - Assay (H₂O₂):30.0 - 32.0 %
- ACS - Color (APHA):= 10
- ACS - Residue after Evaporation:= 20 ppm
- ACS - Free Acid (µeq/g):= 0.6
- Trace Impurities - Aluminum (Al):= 0.2 ppm
- Ammonium (NH₄):= 5 ppm
- Arsenic and Antimony (as As):= 0.01 ppm
- Trace Impurities - Barium (Ba):= 0.01 ppm
- Trace Impurities - Beryllium (Be):= 0.01 ppm
- Trace Impurities - Bismuth (Bi):= 0.01 ppm
- Trace Impurities - Boron (B):= 0.01 ppm
- Trace Impurities - Cadmium (Cd):= 0.01 ppm
- Trace Impurities - Calcium (Ca):= 0.1 ppm
- Chloride (Cl):= 2 ppm
- Trace Impurities - Chromium (Cr):= 0.04 ppm
- Trace Impurities - Cobalt (Co):= 0.01 ppm
- Trace Impurities - Copper (Cu):= 0.01 ppm
- Trace Impurities - Gallium (Ga):= 0.01 ppm
- Trace Impurities - Germanium (Ge):= 0.02 ppm
- Trace Impurities - Gold (Au):= 0.01 ppm
- Heavy Metals (as Pb):= 1 ppm
- Trace Impurities - Iron (Fe):= 0.05 ppm
- Trace Impurities - Lead (Pb):= 0.01 ppm
- Trace Impurities - Lithium (Li):= 0.01 ppm
- Trace Impurities - Magnesium (Mg):= 0.01 ppm
- Trace Impurities - Manganese (Mn):= 0.05 ppm
- Trace Impurities - Mercury (Hg):= 0.001 ppm
- Trace Impurities - Molybdenum (Mo):= 0.01 ppm
- Trace Impurities - Nickel (Ni):= 0.02 ppm
- Nitrate (NO₃):= 2 ppm
- Phosphate (PO₄):= 2 ppm
- Trace Impurities - Potassium (K):= 0.4 ppm
- Trace Impurities - Silver (Ag):= 0.05 ppm
- Trace Impurities - Sodium (Na):= 0.05 ppm
- Trace Impurities - Strontium (Sr):= 0.01 ppm
- ACS - Sulfate (SO₄):= 5 ppm
- Trace Impurities - Thallium (Tl):= 0.05 ppm
- Trace Impurities - Tin (Sn):= 0.5 ppm
- Trace Impurities - Titanium (Ti):= 0.01 ppm
- Trace Impurities - Vanadium (V):= 0.01 ppm
- Trace Impurities - Zinc (Zn):= 0.1 ppm
- Trace Impurities - Zirconium (Zr):= 0.01 ppm
- Macron Fine Chemicals™, Hydrogen peroxide solution 30%, AR Select® ACS for trace metal analysis:
Résultats des tests de spécification
A propos de cet article
:Hydrogène peroxyde
Point d'ébullition:~ 107 °C (1013 hPa)
Point de Fusion:? 0 °C
Densité:1,11 g/cm³ (20 °C)
ADR:5.1,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 150,2 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: -0,41 °C
:Pt Eclair:|Point eclaire:
:M.W. 34,01 g/mol
Point d'ébullition:∼107 °C (1013 hPa)
Densité:1.11 g/cm³ (20 °C)
EINECS:231-765-0
:-0,41
:150,2
ONU:2014
:GHS05
:GHS07
Température d'entreposage:Température ambiante
:14,04798
Point de Fusion:< 0 °C
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 150.2 °C (1013 hPa)
:Melting Pt:
:Melting Pt: < 0 °C
:Flash Pt:
:Flash Pt:
:Density: 1.11 g/cm³ (20 °C)
:M.W. 34.01 g/mol
:Hydrogen peroxide
:34.01
:-0.41
:150.2
:5.1
:II
:1.11
:GHS06
:GHS08
Température d'entreposage:Ambient
:M.O.C. Category N
Customers Who Bought This Also Bought