Dichloromethane ≥99.9% (by GC), ChromAR® for HPLC, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MK487906
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 75-09-2
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Size:1 L
- Supplier No.:4879-06
Specifications
Specification Test Results
- ACS - Assay (CH₂Cl₂) (by GC, Corrected for Water):≥99.9%
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.0003%
- ACS - Titrable Acid (meq/g):≤0.0003
- ACS - Free Halogens:Passes Test
- ACS - Water (H₂O) (by Coulometry):≤0.01%
- Ultraviolet Absorbance (1.00 cm Cell vs. Water) - 235 nm:≤1.0
- Ultraviolet Absorbance (1.00 cm Cell vs. Water) - 240 nm:≤0.12
- Ultraviolet Absorbance (1.00 cm Cell vs. Water) - 250 nm:≤0.10
- Ultraviolet Absorbance (1.00 cm Cell vs. Water) - 260 nm:≤0.04
- Ultraviolet Absorbance (1.00 cm Cell vs. Water) - 340 - 400 nm:≤0.01
- Liquid Chromatography Suitability Absorbance:Passes Test
Specification Test Results
About this item
Synonyms: Methylene chloride
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1,322 g/cm³ (20 °C)
EINECS:200-838-9
ADR:6.1,III
:Management of Change (MOC) category = N
Merck Index:12,06140
UN:1593
:GHS08
:GHS07
Storage Temperature:Température ambiante
Frequently Bought Together