Benzyl alcohol ≥97%, AR®, Macron Fine Chemicals™
# de catalogue MK181604
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 100-51-6
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:500 ml
- Cat. no.:MK181604
- Supplier No.:1816-04
Spécifications
Résultats des tests de spécification
- For Laboratory,Research,or Manufacturing Use:
- Assay (C₆H₅CH₂OH):97.0 - 100.5 %
- Residue on Ignition:≤0.005 %
- Solubility:Passes Test
- Specific Gravity at 25°/25°C:1.042 - 1.047
- Identification:Passes Test
- Refractive Index, n²ºD:1.539 - 1.541
- Acidity:Passes Test
- Nonvolatile Residue:≤0.05 %
- Halogenated Compounds and Halides (as Cl):≤0.03 %
- Benzaldehyde:≤0.20 %
- Appearance (clear, colorless liquid):
- The manufacturer has assurance, based on knowledge of the manufacturing process and controlled handling, shipping and storage of the material, that there is no potential for organic volatile impurities to be present as described in the current U.S.P. In this case unnecessary testing may be avoided.:
Résultats des tests de spécification
A propos de cet article
Synonymes: Phenylmethanol
Point d'ébullition:205 °C (1013 hPa)
Point de Fusion:–15.3 °C
Densité:1.05 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97
:Management of Change (MOC) category = N
Point d'éclair:101 °C
:Boiling Pt:
:Boiling Pt: 205 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –15.3 °C
:Flash Pt:
:Flash Pt: 101 °C
:M.W. 108.14 g/mol
Index Merck:13,01126
EINECS:202-859-9
:-15.3
:GHS07
Température d'entreposage:Ambient
:M.O.C. Category N
Customers Who Bought This Also Bought