Sulfuric acid 95.0-98.0% (by acidimetry) NF, FCC, J.T.Baker®
# de catalogue JT9675-2
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 7664-93-9
MOC : RéglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, clear in a single shipper
- Size:500 ml
- Supplier No.:9675-02
Spécifications
Résultats des tests de spécification
- Food GMP Manufactured Product:-
- GMP Manufactured Product:-
- Meets F.C.C. Requirements:-
- Meets N.F. Requirements:-
- NF - Assay (H₂SO₄) (by acidimetry):95.0 - 98.0 %
- NF - Arsenic (As):≤ 1 ppm
- NF - Chloride (Cl):≤ 0.005 %
- NF - Identification:Passes Test
- NF - Reducing Substances:Passes Test
- NF - Residue on Ignition:≤ 0.005 %
- FCC - Assay (H₂SO₄) (by acidimetry):95.0 - 98.0 %
- FCC - Arsenic (As):≤ 1 ppm
- FCC - Chloride (Cl):≤ 0.005 %
- FCC - Iron (Fe):≤ 0.02 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤ 5 mg/kg
- FCC - Nitrate (NO₃):≤ 10 mg/kg
- FCC - Reducing Substances:Passes Test
- FCC - Selenium (Se):≤ 0.002 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Sulfuric acid 95.0-98.0% (by acidimetry) NF, FCC:
Résultats des tests de spécification
A propos de cet article
Point d'ébullition:~ 335 °C (1013 hPa)
Point de Fusion:3 °C
Densité:1,84 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 330 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: 10,38 °C|Pt Fusion: 3 °C
:Pt Eclair:|Point eclaire:
:M.W. 98,08 g/mol
Index Merck:14,08974
Point d'ébullition:∼335 °C (1013 hPa)
Densité:1.84 g/cm³ (20 °C)
Température d'entreposage:Température ambiante
EINECS:231-639-5
:10,38
:330
ONU:1830
:GHS05
:98.08
:95,0-98,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 330 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 3 °C
:Flash Pt:
:Flash Pt:
:Density: 1.84 g/cm³ (20 °C)
:M.W. 98.08 g/mol
:8
:II
:1.84
:GHS07
:GHS08
Température d'entreposage:Ambient
:M.O.C. Category R
Customers Who Bought This Also Bought