Potassium hydrogen phthalate 99.95-100.05% (dry basis), crystals, BAKER ANALYZED® ACS, J.T.Baker®
# de catalogue JT2958-5
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 877-24-7
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:2.5 kg
- Supplier No.:2958-05
Spécifications
Résultats des tests de spécification
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay (1-KOCOC₆H₄-2-COOH) (dried basis):99.95 - 100.05 %
- Insoluble Matter:≤0.005 %
- pH of 0.05M Solution at 25°C:4.00 - 4.02
- Chlorinated Compounds (as Cl):≤0.003 %
- Sulfur Compounds (as S):≤0.002 %
- Sodium (Na):≤0.005 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- This reagent is satisfactory for use as a pH standard. For use as:
- anacidimetric standard, this material should be lightly crushed and dried for2 hours at 120° C to remove any absorbed moisture.:
- Meets Reagent Specifications for testing USP/NF monographs:
- J.T.Baker®, Potassium hydrogen phthalate 99.95-100.05% (dry basis), crystals, BAKER ANALYZED® ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Potassium phthalate monobasic,Potassium biphthalate,KHP,Phthalic Acid Monopotassium Salt
:Potassium hydrogen phthalate
Point de Fusion:295…300 °C
Densité:1.6362 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,95-100,05
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 295…300 °C
:Flash Pt:
:Flash Pt:
:M.W. 204.22 g/mol
Index Merck:13,07694
Température d'entreposage:Ambient
EINECS:212-889-4
:M.O.C. Category N
Customers Who Bought This Also Bought