Methanol ≥99.8% ACS, VWR Chemicals BDH®
# de catalogue BDH1135-4LG
Fournisseur: VWR International
Product available on GSA Advantage®
Numéro CAS: 67-56-1
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Supplier No.:PL230ZA-4
Spécifications
Résultats des tests de spécification
- Appearance:Clear
- Water (K. F.):Less than 0.10%
- Residue:Less than 0.001% (0.0010 g/125 mL)
- GC Purity:Greater than 99.8%
- Solubility in water:Passes test (Solution should be as clear as an equal volume of water)
- Color (APHA):Less than 10
- Titrable acid:Less than 0.3 meq/g (No more than 0.50 mL of 0.01N sodium hydroxide)
- Titrable base:Less than 0.2 meq/g (No more than 0.40 mL of 0.01N hydrochloric acid)
- Carbonyl compounds (as acetone, formaldehyde, and acetaldehyde):Less than 0.001% each
- Substances darkened by sulfuric acid:Passes test (Solution should be colorless, or have no more color than the acid or methanol before mixing)
- Substances reducing permanganate:Passes test (Pink color should not be entirely discharged)
- VWR Chemicals BDH®, Methanol ≥99.8% ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydroxy methane,Carbinol,Wood alcohol
Point d'ébullition:64,6 °C (1013 hPa)
Point de Fusion:–98 °C
Densité:0,7918 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
Point d'éclair:11 °C (closed cup)
:Point d'ébullition:
:Pt ébul.: 64,6 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: -98 °C
:Point eclaire:
:Pt Eclair: 11 °C (coupe fermée)
:M.W. 32,04 g/mol
Index Merck:13,05984
Température d'entreposage:Température ambiante
EINECS:200-659-6
:-98
:64,6
ONU:1230
:01-2119433307-44
:3
:II
:11
:GHS02
:GHS06
:GHS08
Customers Who Bought This Also Bought