To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Ø:13 mm
- Volume:1.8 ml
- Description:Tube with 3.2% (0.109M) Sodium Citrate Solution
- Length:75 mm
- Cat. no.:95057-169
- Tube Dimensions:13 x 75 mm
- Ring Color:White
- Supplier No.:454322
Specifications
About this item
Designed for use in the examination of coagulation parameters, coagulation tubes contain a buffered tri-sodium citrate solution.
- PET plastic construction is virtually unbreakable
- Contains buffered tri-sodium citrate solution
- Color-coded rings in caps indicate draw volume
Citrate concentrations of either 3.2% (0.109M) or 3.8% (0.129M) are available, with a mixing ratio of one part citrate to nine parts blood.
VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with blue non-ridged pull caps that feature an exterior color-coded ring to indicate the draw volume. A black ring indicates standard tubes and a white ring indicates a tube designed to accommodate pediatric draw. High altitude tubes are recommended for use in locations 5,000ft. or higher above sea level.