ContactUsLinkComponent
Syringe St Cen Ls 5Ml Pk100
HENKE-JECT® 3-Part Disposable Syringes, Air-Tite
# de catalogue 89215-226
Fournisseur:  Air-Tite
Small Business Enterprise
Syringe St Cen Ls 5Ml Pk100
HENKE-JECT® 3-Part Disposable Syringes, Air-Tite
# de catalogue 89215-226
Fournisseur:  Air-Tite
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Sterility:
    Sterile
  • Volume:
    5 ml
  • Cat. no.:
    89215-226
  • For Use With:
    Human, Laboratory and Veterinary Use
  • Type:
    Luer Slip, Centric Tip
  • Supplier No.:
    5050.000VZ
Spécifications
A propos de cet article
HENKE-JECT® syringes are a high quality disposable rubber-piston syringe offering substantial savings.
  • ETO gas sterilized
  • Latex and pyrogen free
A lubricated synthetic rubber piston allows for a smooth draw while a positive stop helps prevent accidental pullout. Bold graduations are easily read and resist wipe off.
Certifications: Meets ISO standards, CE registered.
Packaging: All HENKE-JECT® syringes are packaged within individual sterile strip packs.
Customers Who Bought This Also Bought