ContactUsLinkComponent
OSOM* hCG Combo Test
OSOM® hCG Test Kits, Sekisui Diagnostics
# de catalogue 71001-044
Fournisseur:  Sekisui Diagnostics
Numéro CAS:  
OSOM* hCG Combo Test
OSOM® hCG Test Kits, Sekisui Diagnostics
# de catalogue 71001-044
Fournisseur:  Sekisui Diagnostics
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    hCG Combo Test Kit (Serum and Urine)
  • Cat. no.:
    71001-044
  • Supplier No.:
    124
Spécifications
A propos de cet article
This variety of hCG testing kits and procedures allows users to select the method that best suits the facility in use.
The hCG Urine Test Kit offers an economic and convenient procedure that utilizes a color immunochromatographic dipstick testing method. Easy-to-read two-color results are available in as little as one minute. Test is 99% accurate. The kit includes 50 tests, and maintains an 18-month shelf life from the date of manufacture when stored at room temperature. Sensivity 25 mIU/mL. CLIA-waived.
The Urine Card Pregnancy Test Kit is a traditional physician's office test that utilizes a solid phase, sandwich-format immunochromatographic assay testing method. Tests provide easy-to-read black and white results in 3 minutes. The kit includes 25 tests, and maintains a 24-month shelf life from the date of manufacture when stored at room temperature. Sensivity 20 mIU/mL. CLIA-waived.
The hCG Combo Test Kit is a rapid immunoassay for the qualitative detection of hCG in urine or serum. Designed for use in physician's offices or clinical laboratories, the test utilizes a solid phase, sandwich-format immunochromatographic assay testing method. Urine samples provide results in 3 minutes; serum samples in 5 minutes. The kit includes 25 tests, and maintains a 24-month shelf life from the date of manufacture when stored at room temperature. Sensivity 10 mIU/mL for Serum; 20 mIU/mL for Urine. CLIA-waived for urine.
Urine and serum control sets are sold separately. Urine control set includes one negative and two levels of positive urine controls. Serum control set includes one negative and one positive serum control. Control sets require refrigerated storage.
Customers Who Bought This Also Bought