To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Ampule
- CAS No.:74-83-9
- Size:1 ml
- Formula:CH₃Br
- Description:Bromomethane
- Cat. no.:RK30253
- Supplier No.:30253
Spécifications
A propos de cet article
Solvent packaged in purge-and-trap-grade methanol.
- 2000 μg/mL in P&T methanol, 1 mL/ampoule
Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.