Acetic acid glacial 99.5-100.05% USP, Multi-Compendial, J.T.Baker®
# de catalogue JT9526-33
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 64-19-7
MOC : RéglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, clear, PVC-coated
- Size:2.5 L
- Cat. no.:JT9526-33
- Supplier No.:9526-33
Spécifications
Résultats des tests de spécification
- Meets U.S.P Requirements:—
- Meets J.P. Chemical Specifications:—
- GMP Manufactured Product:—
- Meets B.P. Chemical Specifications:—
- Meets E.P. Chemical Specifications:—
- Bulk Pharmaceutical Chemical:—
- USP - Identification:Passes Test
- USP - Congealing Temperature:= 15.6 °C
- USP - Nonvolatile Residue (USP), mg/20 mL:= 1.0
- USP - Chloride (Cl):Passes Test
- USP - Sulfate (SO₄):Passes Test
- USP - Heavy Metals (as Pb):= 5 ppm
- USP - Readily Oxidizable Substances:Passes Test
- USP - Assay (CH₃COOH):99.5 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance:Passes Test
- EP/BP - Freezing Point:= 14.8 °C
- EP/BP - Heavy Metals (as Pb):= 5 ppm
- EP/BP - Iron (Fe):= 5 ppm
- EP/BP - Chloride (Cl):= 25 ppm
- EP/BP - Sulfate (SO₄):= 50 ppm
- EP/BP - Reducing Substances:Passes Test
- EP/BP - Residue on Evaporation:= 0.01 %
- EP/BP - Assay (CH₃COOH):99.0 - 100.5 %
- JP - Assay (CH₃COOH):99.0 - 101.0 %
- JP - Identification:Passes Test
- JP - Congealing Point:= 14.5 °C
- JP - Nonvolatile Residue, mg/10 mL:= 1.0
- JP - Chloride (Cl):Passes Test
- JP - Sulfate (SO₄):Passes Test
- JP - Heavy Metals (as Pb):= 10 ppm
- JP - Reducing Substances:Passes Test
- Residual Solvents: No Class 1, 2 or 3 residual solvents are used in or:
- produced by the production process.:
- Preserve in tight containers, and store at room temperature:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Acetic acid glacial 99.5-100.05% USP, Multi-Compendial:
Résultats des tests de spécification
A propos de cet article
Synonymes: Vinegar acid,Methanecarboxylic acid
Point d'ébullition:118 °C (1013 hPa)
Point de Fusion:17 °C
Densité:1,05 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
Point d'éclair:38,5 °C
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Fusion: 17 °C
:Pt Eclair:|Point eclaire:
:M.W. 60,05 g/mol
Point d'ébullition:118 °C (1013 hPa)
Densité:1.05 g/cm³ (20 °C)
EINECS:200-580-7
:17
:118
Point d'éclair:38.5 °C
ONU:2789
:01-2119475328-30
:38,5
:GHS02
:GHS05
Température d'entreposage:Température ambiante
:99,5-100,05
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
:8
:II
:GHS07
Température d'entreposage:Ambient
Customers Who Bought This Also Bought