Sodium hydroxide 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
# de catalogue JT5653-2
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 1310-73-2
MOC : With Information"Certificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:PE bottle, narrow mouth
- Size:1 l
- Cat. no.:JT5653-2
- Supplier No.:5653-02
Spécifications
Résultats des tests de spécification
- Normality:0.0195 - 0.0205
- Chloride (Cl):= 1 ppm
- Heavy Metals (as Pb):= 0.5 ppm
- Trace Impurities - Iron (Fe):= 0.5 ppm
- Trace Impurities - Heavy Metals:≤0.5 ppm
- Standardization at 25°C traceable to NIST Standard Reference Material.:
- SRM No…:
Résultats des tests de spécification
A propos de cet article
Synonymes: Caustic soda
Formule:NaOH
Densité:1 g/cm³ (25 °C)
Numéro MDL:MFCD00003548
Numéro CAS:1310-73-2
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Fusion: 323 °C
:Pt Eclair:|Point eclaire:
:M.W. 40 g/mol
EINECS:215-185-5
:323
:1390
Température d'entreposage:Température ambiante
:12,08772
:14,08627
:Storage Temperature: Ambient
:Length
:Width
:Height
:Diameter
:Thickness
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: 1390 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 323 °C
:Flash Pt:
:Flash Pt:
:8
:III
Température d'entreposage:Ambient
:M.O.C. Category NI
Customers Who Bought This Also Bought