Sodium hydrogen carbonate ≥99%, powder USP, Multi-Compendial, J.T.Baker®
# de catalogue JT3510-5
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 144-55-8
MOC : RéglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber, wide mouth
- Size:2.5 kg
- Supplier No.:3510-05
Spécifications
Résultats des tests de spécification
- USP - Identification:Passes Test
- USP - Limit of Sulfur Compounds:= 0.015 %
- USP - Loss on Drying:= 0.25 %
- USP - Insoluble Matter:Passes Test
- USP - Normal Carbonate:Passes Test
- USP - Limit of Ammonia (NH₃):Passes Test
- USP - Chloride (Cl):= 0.015 %
- USP - Arsenic (As):= 2 ppm
- USP - Heavy Metals (as Pb):= 5 ppm
- USP - Assay (NaHCO₃) (dried basis):99.0 - 100.5 %
- USP - Endotoxin Concentration (EU/g):= 2.5
- EP/BP - Assay (NaHCO₃):99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Carbonate (CO₃):Passes Test
- EP/BP - Chloride (Cl):= 150 ppm
- EP/BP - Sulfate (SO₄):= 150 ppm
- EP/BP - Ammonium (NH₄):= 20 ppm
- EP/BP - Arsenic (As):= 2 ppm
- EP/BP - Calcium (Ca):= 100 ppm
- EP/BP - Heavy Metals (as Pb):= 10 ppm
- EP/BP - Iron (Fe):= 20 ppm
- JP - Assay (NaHCO₃):99.0 - 101.0 %
- JP - Identification:Passes Test
- JP - pH (1 in 20):7.9 - 8.4
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):= 0.040 %
- JP - Carbonate (CO₃):Passes Test
- JP - Ammonium (NH₄):Passes Test
- JP - Heavy Metals (as Pb):= 5 ppm
- JP - Arsenic (As):= 2 ppm
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Sodium hydrogen carbonate ≥99%, powder USP, Multi-Compendial:
Résultats des tests de spécification
A propos de cet article
Synonymes: Sodium bicarbonate
Point de Fusion:270 °C
Densité:2,159 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Fusion: 270 °C
:Pt Eclair:|Point eclaire:
:M.W. 84,01 g/mol
Index Merck:13,08655
EINECS:205-633-8
:270
Température d'entreposage:Température ambiante
:12,08726
Densité:2.159 g/cm³ (20 °C)
:99
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 270 °C
:Flash Pt:
:Flash Pt:
:M.W. 84.01 g/mol
Température d'entreposage:Ambient
:M.O.C. Category R
Customers Who Bought This Also Bought