Potassium chloride 99.0-100.5% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
# de catalogue JT3040-1
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 7447-40-7
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:500 g
- Cat. no.:JT3040-1
- Supplier No.:3040-01
Spécifications
Résultats des tests de spécification
- Assay (KCl) (by Ag titrn):99.0 - 100.5%
- Loss on Drying at 105 °C:≤1.0%
- Insoluble Matter:≤0.005%
- pH of 5% Solution at 25 °C:5.4 - 8.6
- Iodide (I):≤0.002%
- Bromide (Br):≤0.01%
- Calcium (Ca):≤0.002%
- Chlorate and Nitrate (as NO₃):≤0.003%
- Magnesium (Mg):≤0.001%
- Nitrogen Compounds (as N):≤0.001%
- Sulfate (SO₄):≤0.001%
- Barium (Ba):Passes Test
- Sodium (Na)(by FES):≤0.005%
- Phosphate (PO₄):≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Arsenic (As):≤1 ppm
- Trace Impurities - Iron (Fe):≤2.0 ppm
- Trace Impurities - Nickel (Ni):≤1.000 ppm
- Meets ACS Reagent Chemical Requirements:≤2.0 ppm
- Meets Reagent Specifications for testing USP/NF monographs:≤1.000 ppm
- J.T.Baker®, Potassium chloride 99.0-100.5% (by argentometric titration), crystals, BAKER ANALYZED® ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Potassiumchloride
Point d'ébullition:1420 °C (1013 hPa)
Point de Fusion:773 °C
Densité:1,98 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 1420 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: 773 °C
:Pt Eclair:|Point eclaire:
:M.W. 74,55 g/mol
Index Merck:14,07621
Point d'ébullition:1420 °C (1013 hPa)
Densité:1.98 g/cm³ (20 °C)
EINECS:231-211-8
:773
:1420
Température d'entreposage:Température ambiante
:12,07783
:99,0-100,5
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 1420 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 773 °C
:Flash Pt:
:Flash Pt:
:M.W. 74.55 g/mol
Température d'entreposage:Ambient
:M.O.C. Category N
Customers Who Bought This Also Bought