Spécifications
- Size:100 g
- Cat. no.:97064-250
- Supplier No.:N605-100G
Spécifications
A propos de cet article
A quality general application agarose that provides good resolution and is cost-effective for high volume users.
- Ideal Agarose for High Throughput
- High Resolution
- Easy Preparation
- Excellent Sensitivity
A quality general application agarose that provides good resolution and is cost-effective for high volume users. It was developed specifically for quick checks of PCR products, plasmids preps, screening and cloning. Features high resolution, easy preparation and set up, a low EEO and excellent sensitivity with low background when stained.