Spécifications
- Size:500 g
- Supplier No.:0710-500G
Spécifications
A propos de cet article
Agarose I™ is for routine nucleic acid analytical/preparative applications.
A standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications. It has a low EEO for shorter electrophoretic runs without compromising resolution. Excellent for blotting techniques and general manipulations.
- All-Purpose, High-Purity Agarose
- Exceptional Band Resolution and Clarity
- Nuclease- and Protease-free
A standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications. It has a low EEO for shorter electrophoretic runs without compromising resolution. Excellent for blotting techniques and general manipulations.