Nichitip Pipette Tips for Nichiryo Pipettes
# de catalogue 89078-454
Fournisseur: Nichiryo America
Spécifications
- Volume:1000 - 10000 µl
- Cat. no.:89078-454
- Sterile:-
- Supplier No.:BMT-Z
Spécifications
A propos de cet article
These tips are made of 100% virgin polypropyleylene.
- High quality
- Free of detectable DNase, RNase, DNA
- Autoclavable
Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
Customers Who Bought This Also Bought