Filter, Inlet, 10um Porosity
# de catalogue 53498-558
Fournisseur: Upchurch Scientific
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Fournisseur: VWR International
$214.30 to $2,349.04
Masterflex® L/S® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Fournisseur: VWR International
$232.26
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
$635.20 to $2,541.50
uTIP™ CleanPak™ Pipette Tip Reload System for Universal Fit Pipettes, Biotix Inc
Fournisseur: Biotix
$81.72 to $326.77
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
$500.19 to $1,790.36