ContactUsLinkComponent
CAPILLARY NUT FOR HP GC PK2
Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
# de catalogue RK21884
Fournisseur:  Restek
CAPILLARY NUT FOR HP GC PK2
Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
# de catalogue RK21884
Fournisseur:  Restek
Spécifications
  • Description:
    Capillary Column Nut
  • Material:
    -
  • For Use With:
    compact Agilent style ferrules
  • Supplier No.:
    21884
Spécifications
A propos de cet article
Capillary column nuts are available in brass or stainless steel.
  • Designed to fit a wider variety of 1/16" ferrules
For use with Agilent 5890/6850/6890/7890 GCs.
Customers Who Bought This Also Bought