Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
# de catalogue RK21884
Fournisseur: Restek
Spécifications
- Description:Capillary Column Nut
- Material:-
- For Use With:compact Agilent style ferrules
- Supplier No.:21884
Spécifications
A propos de cet article
Capillary column nuts are available in brass or stainless steel.
- Designed to fit a wider variety of 1/16" ferrules
For use with Agilent 5890/6850/6890/7890 GCs.
Customers Who Bought This Also Bought